Learn More
Biomatik Corporation Human Amyloid Beta Peptide 1-42, Aβ1-42 Elisa Kit
Supplier:  Biomatik Corporation EKC32566
Encompass_Preferred
Human Amyloid Beta Peptide 1-42, Aβ1-42 Elisa Kit for detecting Amyloid Beta Peptide 1-42, Aβ1-42 96T in serum, plasma, tissue homogenates, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Amyloid beta 42 (ABeta 42); amyloid beta peptide 1-42; Aβ1-42; amyloid βpeptide 1-42. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7311