Learn More
Biomatik Corporation Human Angiopoietin-Like Protein 3, Angptl3 Elisa Kit
Supplier:  Biomatik Corporation EKC32578
Encompass_Preferred
Human Angiopoietin-Like Protein 3, Angptl3 Elisa Kit for detecting Angiopoietin-Like 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y5C1; ANGPTL3. Target Synonyms: ANG 5; ANG-5; ANG5; Angiopoietin 5; Angiopoietin like 3; Angiopoietin related protein 3; Angiopoietin-5; Angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGL3_HUMAN; ANGPT5; ANGPTL3; ANL3; FHBL2; OTTHUMP00000010719; UNQ153/PRO179. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7314