Learn More
Biomatik Corporation Human Angiotensin Converting Enzyme 2, Ace2 Elisa Kit
Supplier:  Biomatik Corporation EKC32592
Encompass_Preferred
Human Angiotensin Converting Enzyme 2, Ace2 Elisa Kit for detecting Angiotensin I Converting Enzyme (Peptidyl-Dipeptidase A) 2 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BYF1; ACE2. Target Synonyms: ACE 2; ACE related carboxypeptidase; ACE-related carboxypeptidase; ACE2; ACE2_HUMAN; ACEH; Angiotensin converting enzyme 2; Angiotensin converting enzyme homolog; Angiotensin converting enzyme like protein; Angiotensin I Converting Enzyme (peptidyl dipeptidase A) 2; Angiotensin I converting enzyme 2; Angiotensin-converting enzyme homolog; DKFZP434A014; EC 3.4.17; metalloprotease MPROT 15; Metalloprotease MPROT15; OTTHUMP00000022963; Processed angiotensin-converting enzyme 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7316