Learn More
Biomatik Corporation Hydroxyproline (Hyp) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04857
Encompass_Preferred
Hydroxyproline (Hyp) ELISA Kit for detecting Hyp in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 61.7-5,000ng/ml. Sensitivity: 28.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: (2S,4R)-4-Hydroxyproline; L-Hydroxyproline (C5H9O3N). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2762