Learn More
Biomatik Corporation Human Annexin Iii (Anx-Iii) Elisa Kit
Supplier:  Biomatik Corporation EKC32602
Encompass_Preferred
Human Annexin Iii (Anx-Iii) Elisa Kit for detecting Annexin A3 in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12429; ANXA3. Target Synonyms: 2-cyclic phosphate 2-phosphohydrolase; 35-alpha calcimedin; Annexin A3; Annexin III; Annexin-3; ANX3; ANXA 3; Anxa3; ANXA3_HUMAN; Calcimedin 35 alpha; Inositol 1 2 cyclic phosphate 2 phosphohydrolase; Inositol 1; Lipocortin III; OTTHUMP00000160701; OTTHUMP00000219237; OTTHUMP00000219238; PAP III; PAP-III; Placental anticoagulant protein III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7318