Learn More
Biomatik Corporation Human Apolipoprotein A-Iv (Apoa4) Elisa Kit
Supplier:  Biomatik Corporation EKC32757
Encompass_Preferred
Human Apolipoprotein A-Iv (Apoa4) Elisa Kit for detecting Apolipoprotein A-Iv(Apoa4) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.041-30ng/ml. Sensitivity: 0.041ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06727. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7326