Learn More
Biomatik Corporation Human Apolipoprotein L1 (Apol1) Elisa Kit
Supplier:  Biomatik Corporation EKC32764
Encompass_Preferred
Human Apolipoprotein L1 (Apol1) Elisa Kit for detecting Apolipoprotein L, 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: O14791; APOL1. Target Synonyms: APO L; Apo-L; ApoL; APOL I; ApoL-I; APOL1; APOL1_HUMAN; APOLI; Apolipoprotein L; Apolipoprotein L I; Apolipoprotein L-I; Apolipoprotein L1; FSGS4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7328