Learn More
Biomatik Corporation Human Aspartate Aminotransferase, Ast Elisa Kit
Supplier:  Biomatik Corporation EKC32800
Encompass_Preferred
Human Aspartate Aminotransferase, Ast Elisa Kit for detecting Glutamic-Oxaloacetic Transaminase 1, Soluble (Aspartate Aminotransferase 1) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20mU/ml. Sensitivity: 0.078mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17174; GOT1. Target Synonyms: AATC_HUMAN; Aspartate aminotransferase 1; Aspartate aminotransferase; Aspartate aminotransferase cytoplasmic; Aspartate aminotransferase cytosolic; ASTQTL1; cAspAT; cCAT; cysteine aminotransferase; cytoplasmic; cysteine transaminase; cytoplasmic; cytoplasmic; ec 2.6.1.1; GIG 18; GIG18; Glutamate oxaloacetate transaminase 1; Glutamate oxaloacetate transaminase soluble; Glutamic oxaloacetic transaminase 1 soluble; GOT 1; GOT1; Growth inhibiting protein 18; SGOT; Transaminase A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7331