Learn More
Biomatik Corporation Human Atrial Natriuretic Peptide (Anp) Elisa Kit
Supplier:  Biomatik Corporation EKC32814
Encompass_Preferred
Human Atrial Natriuretic Peptide (Anp) Elisa Kit for detecting Natriuretic Peptide Precursor A in serum, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01160; NPPA. Target Synonyms: ANF; ANF_HUMAN; ANP; ATFB6; Atrial natriuretic factor; Atrial natriuretic peptide; Atriopeptin; Cardiodilatin; Cardiodilatin related peptide; Cardiodilatin-related peptide; Cardionatrin; CDD ANF; CDD; CDD-ANF; CDP; Natriuretic peptide A; Natriuretic peptide precursor A; NPPA; PND; Prepronatriodilatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7334