Learn More
Biomatik Corporation Human Bradykinin, Bk Elisa Kit
Supplier:  Biomatik Corporation EKC32908
Encompass_Preferred
Human Bradykinin, Bk Elisa Kit for detecting Bradykinin, Bk in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20pg/ml. Sensitivity: 0.078pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7347