Learn More
Biomatik Corporation Human Immunoglobulin A (IgA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04878
Encompass_Preferred
The Human Immunoglobulin A (IgA) ELISA Kit allows for quantitative determination of Immunoglobulin A in Serum, Plasma, Saliva and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.14-100 ng/mL. Sensitivity: 0.07 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Immunoglobulin A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2773