Learn More
Biomatik Corporation Human Cd9 Antigen (Cd9) Elisa Kit
Supplier:  Biomatik Corporation EKC33082
Encompass_Preferred
Human Cd9 Antigen (Cd9) Elisa Kit for detecting Cd9 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21926; CD9. Target Synonyms: CD9; MIC3; TSPAN29; GIG2; CD9 antigen; 5H9 antigen; Cell growth-inhibiting gene 2 protein; Leukocyte antigen MIC3; Motility-related protein; MRP-1; Tetraspanin-29; Tspan-29; p24; CD antigen CD9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7359