Learn More
Biomatik Corporation Pig Immunoglobulin A (IgA) ELISA Kit, 96 tests
Catalog No. 501502775
Encompass_Preferred
Pig Immunoglobulin A (IgA) ELISA Kit for detecting IgA in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.2-2,000pg/ml. Sensitivity: 14.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: K7ZRK0. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2775 Supplier Biomatik Corporation Supplier No. EKU04880
Pig Immunoglobulin A (IgA) ELISA Kit for detecting IgA in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 31.2-2,000pg/ml. Sensitivity: 14.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: K7ZRK0. Restrictions: For Research Use Only. Not for use in diagnostic procedures.