Learn More
Biomatik Corporation Human Clara Cell Protein, Cc16 Elisa Kit
Supplier:  Biomatik Corporation EKC33141
Encompass_Preferred
Human Clara Cell Protein, Cc16 Elisa Kit for detecting Clara Cell Protein, Cc16 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11684; SCGB1A1. Target Synonyms: Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell phospholipid binding protein; Clara cell phospholipid-binding protein; Clara cell specific 10 kD protein; Clara cells 10 kDa secretory protein; OTTHUMP00000236107; SCGB1A1; Secretoglobin family 1A member 1; Secretoglobin; family 1A; member 1 (uteroglobin); UG; UGB; UP-1; UP1; Urinary protein 1; Urine protein 1; UTER_HUMAN; Uteroglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7361