Learn More
Biomatik Corporation Rat Immunoglobulin A (IgA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04881
Encompass_Preferred
Rat Immunoglobulin A (IgA) ELISA Kit for detecting IgA in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.0ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20760. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2776