Learn More
Biomatik Corporation Human Cxc-Chemokine Receptor 4 (Cxcr4) Elisa Kit
Supplier:  Biomatik Corporation EKC33337
Encompass_Preferred
Human Cxc-Chemokine Receptor 4 (Cxcr4) Elisa Kit for detecting Chemokine (C-X-C Motif) Receptor 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P61073; CXCR4. Target Synonyms: CXCR4; C-X-C chemokine receptor type 4; CXC-R4; CXCR-4; FB22; Fusin; HM89; LCR1; Leukocyte-derived seven transmembrane domain receptor; LESTR; Lipopolysaccharide-associated protein 3; LAP-3; LPS-associated protein 3; NPYRL; Stromal cell-derived factor 1 receptor; SDF-1 receptor; CD antigen CD184. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7391