Learn More
Biomatik Corporation Human Immunoglobulin G (IgG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04895
Encompass_Preferred
The Human Immunoglobulin G (IgG) ELISA Kit allows for quantitative determination of Immunoglobulin G in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.23-100 ug/mL. Sensitivity: 0.51 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Immunoglobulin G and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2782