Learn More
Biomatik Corporation Human Inositol 1, 4, 5, -Trisphosphate (Ip3) Elisa Kit
Supplier:  Biomatik Corporation EKC34135
Encompass_Preferred
Human Inositol 1, 4, 5, -Trisphosphate (Ip3) Elisa Kit for detecting Inositol 1, 4, 5, -Trisphosphate(Ip3) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 5-1000pg/ml. Sensitivity: 2.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7443