Learn More
Biomatik Corporation Human Insulin-Like Growth Factors Binding Protein 2, Igfbp-2 Elisa Kit
Supplier:  Biomatik Corporation EKC34157
Encompass_Preferred
Human Insulin-Like Growth Factors Binding Protein 2, Igfbp-2 Elisa Kit for detecting Insulin-Like Growth Factor Binding Protein 2, 36Kda in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18065; IGFBP2. Target Synonyms: BP 2; BP2; IBP 2; IBP-2; IBP2; IBP2_HUMAN; IGF binding protein 2; IGF BP53; IGF-binding protein 2; IGFBP 2; IGFBP-2; IGFBP2; IGFBP53; Insulin like growth factor binding protein 2 36kDa; Insulin like growth factor binding protein 2; Insulin like growth factor-binding protein 2 precursor; Insulin-like growth factor-binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7448