Learn More
Biomatik Corporation Mouse Immunoglobulin G1 (IgG1) ELISA Kit, 96 tests
Catalog No. 501502784
Encompass_Preferred
Mouse Immunoglobulin G1 (IgG1) ELISA Kit for detecting IgG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 246.9-20,000ng/ml. Sensitivity: 89.9ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A075B5P4. Target Synonyms: IGHG1; Ig Gamma-1 Chain C Region; Immunoglobulin Heavy Constant Gamma 1; G1m Marker; Immunoglobulin Gm1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2784 Supplier Biomatik Corporation Supplier No. EKU04901
Mouse Immunoglobulin G1 (IgG1) ELISA Kit for detecting IgG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 246.9-20,000ng/ml. Sensitivity: 89.9ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A075B5P4. Target Synonyms: IGHG1; Ig Gamma-1 Chain C Region; Immunoglobulin Heavy Constant Gamma 1; G1m Marker; Immunoglobulin Gm1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.