Learn More
Biomatik Corporation Human Neutrophil Peptide 1-3, Hnp1-3 Elisa Kit
Supplier:  Biomatik Corporation EKC34816
Encompass_Preferred
Human Neutrophil Peptide 1-3, Hnp1-3 Elisa Kit for detecting Neutrophil Peptide 1-3, Hnp1-3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7496