Learn More
Biomatik Corporation Human Perforin-1 (Prf1/Pfp) Elisa Kit
Supplier:  Biomatik Corporation EKC34955
Encompass_Preferred
Human Perforin-1 (Prf1/Pfp) Elisa Kit for detecting Perforin 1 (Pore Forming Protein) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14222; PRF1. Target Synonyms: Cytolysin; FLH2; HPLH2; Lymphocyte pore-forming protein; P1; PERF_HUMAN; perforin 1 (pore forming protein); Perforin 1; Perforin-1; PFP; PGFL; PIGF; PIGF-2; PLGF; Pore forming protein; prf1; SHGC-10760. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7510