Learn More
Biomatik Corporation Human Pyruvate Kinase, M2-Pk Elisa Kit
Supplier:  Biomatik Corporation EKC35325
Encompass_Preferred
Human Pyruvate Kinase, M2-Pk Elisa Kit for detecting Pyruvate Kinase, Muscle in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60U/ml. Sensitivity: 0.225u/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14618; PKM. Target Synonyms: CTHBP; Cytosolic thyroid hormone-binding protein; KPYM_HUMAN; OIP-3; Opa-interacting protein 3; p58; pkm; PKM1; PKM2; Pyruvate kinase 2/3; Pyruvate kinase muscle isozyme; Pyruvate kinase PKM; THBP1; Thyroid hormone-binding protein 1; Tumor M2-PK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7536