Learn More
Biomatik Corporation Human Inducible T-Cell Co Stimulator (ICOS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04920
Encompass_Preferred
The Human Inducible T-Cell Co Stimulator (ICOS) ELISA Kit allows for quantitative determination inducible T-Cell Co Stimulator in Serum, Plasma, Urine, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y6W8. Cross Activity: No significant cross-reactivity or interference between Inducible T-Cell Co Stimulator and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2790