Learn More
Biomatik Corporation Monkey Complement 4 (C4) Elisa Kit
Supplier:  Biomatik Corporation EKC36147
Encompass_Preferred
Monkey Complement 4 (C4) Elisa Kit for detecting Complement 4(C4) in serum, plasma, cell culture supernates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.41-300ng/ml. Sensitivity: 0.41ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7561