Learn More
Biomatik Corporation Mouse Inhibin A (INHA) ELISA Kit, 96 tests
Catalog No. 501502792
Encompass_Preferred
Mouse Inhibin Alpha (INHa) ELISA Kit for detecting INHa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04997. Gene ID: 16322. Target Synonyms: INH-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2792 Supplier Biomatik Corporation Supplier No. EKU04923
Mouse Inhibin Alpha (INHa) ELISA Kit for detecting INHa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04997. Gene ID: 16322. Target Synonyms: INH-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.