Learn More
Biomatik Corporation Rat Inhibin A (INHA) ELISA Kit, 96 tests
Catalog No. 501502793
Encompass_Preferred
Rat Inhibin Alpha (INHa) ELISA Kit for detecting INHa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17490. Gene ID: 24504. Target Synonyms: INH-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2793 Supplier Biomatik Corporation Supplier No. EKU04925
Rat Inhibin Alpha (INHa) ELISA Kit for detecting INHa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17490. Gene ID: 24504. Target Synonyms: INH-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.