Learn More
Biomatik Corporation Mouse Chitinase-3-Like Protein 1 (Chi3L1) Elisa Kit
Supplier:  Biomatik Corporation EKC36539
Encompass_Preferred
Mouse Chitinase-3-Like Protein 1 (Chi3L1) Elisa Kit for detecting Chitinase 3-Like 1 (Cartilage Glycoprotein-39) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61362; Chi3l1. Target Synonyms: Chi3l1; Brp39; Chil1; Chitinase-3-like protein 1; BRP39 protein; Breast regression protein 39; Cartilage glycoprotein 39; CGP-39; GP-39. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7582