Learn More
Biomatik Corporation Mouse Fetuin A ELISA Kit
Supplier:  Biomatik Corporation EKC36851
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25 ng/mL-400 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P29699. Target Synonyms: A2HS, AHS, FETUA, HSGA, fetuin-A. Restriction: For Research Use Only.
Catalog No. 50-228-7593