Learn More
Biomatik Corporation Mouse Tissue Inhibitors Of Metalloproteinase 2, Timp-2 Elisa Kit
Supplier:  Biomatik Corporation EKC37930
Encompass_Preferred
Mouse Tissue Inhibitors Of Metalloproteinase 2, Timp-2 Elisa Kit for detecting Timp Metallopeptidase Inhibitor 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P25785; Timp2. Target Synonyms: Timp2; Timp-2; Metalloproteinase inhibitor 2; Tissue inhibitor of metalloproteinases 2; TIMP-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7624