Learn More
Biomatik Corporation Mouse Inhibin Beta E (INHbE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04942
Encompass_Preferred
Mouse Inhibin Beta E (INHbE) ELISA Kit for detecting INHbE in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.09-250pg/ml. Sensitivity: 1.07pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O08717. Gene ID: 16326. Target Synonyms: INH-bE; Activin beta-E chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2797