Learn More
Biomatik Corporation Pig Insulin,INS ELISA Kit
Supplier:  Biomatik Corporation EKC38222
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 2 μIU/mL-400 μIU/mL. Sensitivity: 1 μIU/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01315. Target Synonyms: ILPR, IRDN, OTTHUMP00000011162|OTTHUMP00000196038|proinsulin. Restriction: For Research Use Only.
Catalog No. 50-228-7634