Learn More
Biomatik Corporation Human Inosine 5'-Monophosphate Dehydrogenase 2 (IMPDH2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04947
Encompass_Preferred
The Human Inosine 5'-Monophosphate Dehydrogenase 2 (IMPDH2) ELISA Kit allows for quantitative determination inosine 5'-Monophosphate Dehydrogenase 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12268. Cross Activity: No significant cross-reactivity or interference between Inosine 5'-Monophosphate Dehydrogenase 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2798