Learn More
Biomatik Corporation Rat cluster of differentiation 4,CD4 ELISA Kit
Supplier:  Biomatik Corporation EKC38847
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.78 U/mL-50 U/mL. Sensitivity: 0.195 U/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P05540. Target Synonyms: CD4mut, CD4 antigen|CD4 antigen (p55)|CD4 receptor. Restriction: For Research Use Only.
Catalog No. 50-228-7651