Learn More
Biomatik Corporation Guinea Pig Insulin (INS) ELISA Kit, 96 tests
Catalog No. 501502800
Encompass_Preferred
The Guinea Pig Insulin (INS) ELISA Kit allows for quantitative determination insulin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Cavia (Guinea pig). Detection Range: 123.5-10000 pg/mL. Sensitivity: 51.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01329. Cross Activity: No significant cross-reactivity or interference between Insulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2800 Supplier Biomatik Corporation Supplier No. EKU04955
The Guinea Pig Insulin (INS) ELISA Kit allows for quantitative determination insulin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Cavia (Guinea pig). Detection Range: 123.5-10000 pg/mL. Sensitivity: 51.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01329. Cross Activity: No significant cross-reactivity or interference between Insulin and analogues was observed. Restriction: For Research Use Only.