Learn More
Biomatik Corporation Human Insulin (INS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04956
Encompass_Preferred
The Human Insulin (INS) ELISA Kit allows for quantitative determination insulin in Serum, Plasma, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 61.7-5000 pg/mL. Sensitivity: 25.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01308. Cross Activity: No significant cross-reactivity or interference between Insulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2801