Learn More
Biomatik Corporation Human Acidic Fibroblast Growth Factor, Afgf/Fgf-1 Elisa Kit
Supplier:  Biomatik Corporation EKC40041
Encompass_Preferred
Human Acidic Fibroblast Growth Factor, Afgf/Fgf-1 Elisa Kit for detecting Fibroblast Growth Factor 1 (Acidic) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05230; FGF1. Target Synonyms: Acidic fibroblast growth factor; aFGF; Beta endothelial cell growth factor; Beta-endothelial cell growth factor; ECGF; ECGF beta; ECGF-beta; ECGFA; ECGFB; Endothelial Cell Growth Factor alpha; Endothelial Cell Growth Factor beta; FGF 1; FGF alpha; Fgf1; FGF1_HUMAN; FGFA; Fibroblast Growth Factor 1 Acidic; Fibroblast growth factor 1; GLIO703; HBGF 1; HBGF-1; HBGF1; Heparin binding growth factor 1; Heparin binding growth factor 1 precursor; Heparin-binding growth factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7694