Learn More
Biomatik Corporation Mouse Insulin (INS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04957
Encompass_Preferred
Mouse Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.65pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01326. Gene ID: 16334. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2802