Learn More
Biomatik Corporation Human Interleukin 11, Il-11 Elisa Kit
Supplier:  Biomatik Corporation EKC40067
Encompass_Preferred
Human Interleukin 11, Il-11 Elisa Kit for detecting Interleukin 11 in serum, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20809; IL11. Target Synonyms: Adipogenesis inhibitory factor; AGIF; IL 11; IL-11; Il11; IL11_HUMAN; Interleukin 11; Interleukin-11; Oprelvekin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7699