Learn More
Biomatik Corporation Pig Immunoglobulin M,IgM ELISA Kit
Supplier:  Biomatik Corporation EKC40225
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Sus scrofa (Pig). Detection Range: 0.039 μg/mL-10 μg/mL. Sensitivity: 0.039 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7715