Learn More
Biomatik Corporation Dog Insulin Like Growth Factor 1 (IGF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04964
Encompass_Preferred
The Dog Insulin Like Growth Factor 1 (IGF1) ELISA Kit allows for quantitative determination insulin Like Growth Factor 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 78-5000 pg/mL. Sensitivity: 31 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P33712. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2806