Learn More
Biomatik Corporation Human Keratinocyte Growth Factor, Kgf Elisa Kit
Supplier:  Biomatik Corporation EKC40541
Encompass_Preferred
Human Keratinocyte Growth Factor, Kgf Elisa Kit for detecting Fibroblast Growth Factor 7 (Keratinocyte Growth Factor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21781; FGF7. Target Synonyms: FGF 7; FGF-7; Fgf7; FGF7_HUMAN; Fibroblast growth factor 7; HBGF 7; HBGF-7; HBGF7; Heparin binding growth factor 7; Heparin-binding growth factor 7; Keratinocyte growth factor; KGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7750