Learn More
Biomatik Corporation Human Insulin Like Growth Factor 1 (IGF1) ELISA Kit, 96 tests
Catalog No. 501502808
Encompass_Preferred
The Human Insulin Like Growth Factor 1 (IGF1) ELISA Kit allows for quantitative determination insulin Like Growth Factor 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05019. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2808 Supplier Biomatik Corporation Supplier No. EKU04967
The Human Insulin Like Growth Factor 1 (IGF1) ELISA Kit allows for quantitative determination insulin Like Growth Factor 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05019. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor 1 and analogues was observed. Restriction: For Research Use Only.