Learn More
Biomatik Corporation Human Thymus Activation Regulated Chemokine, Tarc Elisa Kit
Supplier:  Biomatik Corporation EKC40594
Encompass_Preferred
Human Thymus Activation Regulated Chemokine, Tarc Elisa Kit for detecting Chemokine (C-C Motif) Ligand 17 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92583; CCL17. Target Synonyms: A-152E5.3; A152E53; ABCD 2; ABCD2; C-C motif chemokine 17; CC chemokine TARC; CCL17; CCL17_HUMAN; Chemokine CC Motif Ligand 17; MGC138271; MGC138273; SCYA17; Small Inducible Cytokine A17; Small Inducible Cytokine A17 Precursor; Small Inducible Cytokine Subfamily A (Cys Cys); Small Inducible Cytokine Subfamily A (Cys Cys) Member 17; Small-inducible cytokine A17; T Cell Directed CC Chemokine; Thymus and activation regulated chemokine; Thymus and activation-regulated chemokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7760