Learn More
Biomatik Corporation Escherichia Coli Protein (E. Coli P) Elisa Kit
Supplier:  Biomatik Corporation EKC40798
Encompass_Preferred
Escherichia Coli Protein (E. Coli P) Elisa Kit for detecting Escherichia Coli Protein (E. coli P) in serum, plasma, tissue homogenates. Species Reactivity: Escherichia Coli. Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7787