Learn More
Biomatik Corporation Rabbit Insulin Like Growth Factor 1 (IGF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04971
Encompass_Preferred
Rabbit Insulin Like Growth Factor 1 (IGF1) ELISA Kit for detecting IGF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q95222. Gene ID: 100008668. Target Synonyms: IGFI; IGF1A; IBP1; MGF; Somatomedin C; Mechano Growth Gactor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2812