Learn More
Biomatik Corporation Human Heat Shock Protein 90Α (Hsp-90Α) Elisa Kit
Supplier:  Biomatik Corporation EKC41013
Encompass_Preferred
Human Heat Shock Protein 90Α (Hsp-90Α) Elisa Kit for detecting Heat Shock Protein 90Α (Hsp-90Α) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07900; HSP90AA1. Target Synonyms: Heat shock 86 kDa; Heat shock protein 90kDa alpha cytosolic class A member 1; Heat shock protein 90kDa alpha cytosolic class B member 1; Heat shock protein HSP 90 alpha; Heat shock protein HSP 90 beta; Heat shock protein HSP 90-alpha; HS90A_HUMAN; HSP 84; HSP 86; Hsp 90; HSP86; HSP90A; HSP90AA1; HSP90AB1; HSP90B; HSPC1; HSPC2; HSPCAL1; HSPCAL4; Renal carcinoma antigen NY-REN-38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7812