Learn More
Biomatik Corporation Human Progranulin (Pgrn) Elisa Kit
Supplier:  Biomatik Corporation EKC41165
Encompass_Preferred
Human Progranulin (Pgrn) Elisa Kit for detecting Progranulin(Pgrn) in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.7-300ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7829