Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04987
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 1 (IGFBP1) ELISA Kit for detecting IGFBP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21743. Gene ID: 25685. Target Synonyms: IBP1; AFBP; IGF-BP25; PP12; Placental Protein 12; Amniotic Fluid Binding Protein; Alpha-Pregnancy-Associated Endometrial Globulin; Growth Hormone Independent-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2816