Learn More
Biomatik Corporation Human Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04990
Encompass_Preferred
The Human Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit allows for quantitative determination insulin Like Growth Factor Binding Protein 2 in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 27.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18065. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor Binding Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2817